Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FEN-1 Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI1F3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270716AF700

Catalog No. N270716A700


Add to cart

Description

Description

FEN-1 Monoclonal antibody specifically detects FEN-1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

FEN-1
OTI1F3
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
FEN1
Recombinant protein fragment corresponding to amino acids 117-380 of human FEN-1 (NP_004102) produced in E.coli.
Immunogen affinity purified
Base Excision Repair, Cancer, DNA Repair, Editing and Processing Endonucleases
Primary
2237.0
Immunohistochemistry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
EC 3.1, FEN-1hFEN-1, flap structure-specific endonuclease 1DNase IV, Maturation factor 1, maturation factor-1, MF1flap endonuclease 1, RAD2
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit