Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fenbendazole sulfone, Honeywell Fluka™

Click to view available options
Quantity:
10 mg
54029-20-8
C15H13N3O4S
331.35
MFCD00797921
54029-20-8
C15H13N3O4S
331.35
MFCD00797921

Analytical standard, reference material

Ensure you have the information you need to quickly start your analysis, and pass relevant audits:

  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
Beilstein 703211
Physical Form Crystalline powder
Grade Analytical Standard
Packaging Glass Bottle
Quantity 10 mg
For Use With (Application) ANTHELMINTICS AND FLUKICIDES

P280

H302+H315+H317

Warning

Recommended Storage : Room Temp