Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fenbendazole, USP, 98-101%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
43210-67-9
C15H13N3O2S
299.35
HDDSHPAODJUKPD-UHFFFAOYSA-N
methyl N-[6-(phenylsulfanyl)-1H-1,3-benzodiazol-2-yl]carbamate
COC(=O)NC1=NC2=CC=C(SC3=CC=CC=C3)C=C2N1
43210-67-9
C15H13N3O2S
299.35
HDDSHPAODJUKPD-UHFFFAOYSA-N
methyl N-[6-(phenylsulfanyl)-1H-1,3-benzodiazol-2-yl]carbamate
COC(=O)NC1=NC2=CC=C(SC3=CC=CC=C3)C=C2N1

F1359, 43210-67-9, C15H13N3O2S

Spectrum™ Fenbendazole, USP is used as a broad spectrum benzimidazole anthelmintic that guards against gastrointestinal parasites in animals, mostly cattle, sheep, dogs, cats and rabbits. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.003
Quantity 5 g
Loss on Drying 0.01
CAS Max % 1
Percent Purity 98 to 101%