Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fenbufen, MilliporeSigma™ Supelco™

Fenbufen belongs to the class of non-steroidal anti-inflammatory drugs, widely used as an antipyretic and analgesic in medical applications. Its mode of action involves the inhibition of cyclooxygenase enzyme and thereby prevents the synthesis of certain prostaglandins.

$104.70 - $176.40

Specifications

CAS 36330-85-5
Grade Analytical Standard
Linear Formula C6H5C6H4 CO(CH2)2 CO2 H
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-8630
View Documents
MilliporeSigma Supelco
F875510G
10 g
Each for $176.40
Add to cart
 
11-101-9113
View Documents
MilliporeSigma Supelco
F87555G
5 g
Each for $104.70
Add to cart
 
Description

Description

Featured Industries: Pharmaceutical (small molecule)
Specifications

Specifications

36330-85-5
Analytical Standard
C6H5C6H4 CO(CH2)2 CO2 H
Neat
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit