Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FER Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5121237

This item is not returnable.
View return policy
Description
Positive test controls include: HeLa, HT-1080, SW480, LO2, NIH/3T3, Mouse liver, Mouse heart, Rat brain. The target is usually found in the following locations: Cell junction, Cell membrane, Cell projection, Cytoplasm, Cytoplasmic side, Membrane, Nucleus, Peripheral membrane protein, cell cortex, cytoskeleton.
FER Polyclonal antibody specifically detects FER in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
FER | |
Polyclonal | |
Unconjugated | |
FER | |
AV082135; C330004K01Rik; c-FER; EC 2.7.10.2; Feline encephalitis virus-related kinase FER; FER; fer (fms/fps related) protein kinase; fer (fms/fps related) protein kinase, testis specific 2; fer (fps/fes related) tyrosine kinase; FER tyrosine kinase; Fert; Fert2; Flk; Flk_retired; Fujinami poultry sarcoma/Feline sarcoma-related protein Fer; p47; p94-Fer; phosphoprotein NCP94; PPP1R74; protein phosphatase 1, regulatory subunit 74; proto-oncogene c-Fer; proto-oncogene tyrosine-protein kinase Fer; TYK3; Tyrosine kinase 3; tyrosine protein kinase FLK; tyrosine-protein kinase Fer; Tyrosine-protein kinase FLK | |
Recombinant fusion protein containing a sequence corresponding to amino acids 200-450 of human FER (NP_005237.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
1.9 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P09760, P16591, P70451 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14158, 2241, 301737 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title