Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FER Mouse anti-Mouse, Clone: 1487CT794.8.50, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524734

Catalog No. PIMA524734

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Protein kinases constitute a large superfamily of enzymes with key regulatory functions in nearly all signal transmission processes of eukaryotic cells (1). FER is found in the cytoplasm and in the nucleus, where it was associated with the chromatin fraction. Its ubiquitous expression and its subcellular location indicate that it may be involved in key regulatory processes (2). The group IV cytoplasmic protein-tyrosine kinase Fer has been linked to cellular signaling responses to many different stimuli, including growth factors and cytokines. Data suggest a biological role for Fer in regulation of leukocyte recruitment during the innate immune response (3).
Specifications

Specifications

FER
Monoclonal
0.89 mg/mL
PBS with 0.09% sodium azide
P70451
Tyrosine-protein kinase Fer, Proto-oncogene c-Fer, p94-Fer, Fer, Fert2
Mouse
IgG2a, kappa
400 μL
-20°C
Primary
14158
Immunocytochemistry, Immunofluorescence, Western Blot
1487CT794.8.50
Unconjugated
Fer
Liquid
Fer
Mouse Fer recombit protein
Protein G
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit