Learn More
Sklar InstrumentsSupplier Diversity Partner Small Ferguson Gallstone Scoop, Sklar, 9-1/2 in., Silver Plated, Double End. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 321035
Small Ferguson Gallstone Scoop, Sklar, 9-1/2 in., Silver Plated, Double End. Sklar's Ferguson Gallstone Scoop is a rigid, double-ended gall bladder instrument with spoon shaped tips used for removing gallstones from the gall bladder. It is avalable in three different sizes.
Catalog No. 50-143-8177