Learn More
Thermo Scientific™ BCIP-T
Enhance blue color development with BCIP-T, a chromogenic substrate for alkaline phosphatase used in conjunction with the oxidant NBT.
Manufacturer:  Thermo Scientific™ R0821
 View more versions of this product
Catalog No. FERR0821
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Alkaline phosphatase catalyses the removal of a phosphate group from BCIP-T, generating a product that oxidizes and dimerizes to dibromodichloro indigo. The reducing equivalents produced during the dimerization reaction reduce NBT to an insoluble purple dye, diformazan.
Alkaline phosphatase catalyses the removal of a phosphate group from BCIP-T, generating a product that oxidizes and dimerizes to dibromodichloro indigo. The reducing equivalents produced during the dimerization reaction reduce NBT to an insoluble purple dye, diformazan (1)
Recommended for:
Detection of alkaline phosphatase activity in:
  • Southern blotting
  • Western blotting
  • Northern blotting
  • dot/slot blots
  • plaque and colony screening
Note:
Preparation of a 50mg/mL stock solution in dimethylformamide is recommended.
Specifications
Specifications
Detection of alkaline phosphatase activity in:; Southern blotting Western blotting Northern blotting dot/slot blots plaque and colony screening;
BCIP-T
1g