Learn More
Thermo Scientific™ Biotin DecaLabel DNA Labeling Kit
GSA_VA
Efficiently synthesize biotin-labeled DNA probes with the Thermo Scientific™ Biotin DecaLabel DNA Labeling Kit.
$197.00 - $447.50
Products 2
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
FERK0651
GSA_VA
View Documents Promotion Details
Thermo Scientific™
K0651
10
Each for $197.00
Add to cart
FERK0652
View Documents Promotion Details
Thermo Scientific™
K0652
30
Each for $447.50
Add to cart
Description
Description
Biotin DecaLabel DNA Labeling Kit is an advanced system for the efficient synthesis of biotin-labeled DNA probes, based on an improved random-primed labeling method developed by Feinberg and Vogelstein. The primary improvement over traditional random-primed method involves the use of random decamers instead of hexamers, ensuring more efficient annealing with DNA at 37°C. Klenow Fragment, exo-, included in the kit, is genetically engineered enzyme with no detectable exonuclease activity. The enzyme does not degrade the labeled probe during reaction, which results in a high labeling yield even with low amounts of template. You can uniformly label any length DNA fragments.
Biotin-labeled DNA is detected with the Biotin Chromogenic Detection Kit or conventional biotin-avidin or biotin-streptavidin detection systems.
Highlights
  • Non-radioactive labeling of DNA
  • Efficient priming of labeling reactions with random decamers
  • High yields with Klenow Fragment, exo-: no degradation of a labeled probe during reaction
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.