Learn More
Thermo Scientific IPTG
Induce the expression of genes under the control of lac operon in cloning and protein expression procedures.
Manufacturer: Thermo Scientific R0391
Description
IPTG (isopropyl-beta-D-thiogalactopyranoside) is a highly stable synthetic analog of lactose. It inactivates the lac repressor and induces synthesis of beta-galactosidase, an enzyme that promotes lactose utilization. IPTG is used to induce expression of cloned genes under control of the lac operon . It is used in conjunction with X-Gal (#R0941) to determine the lac phenotype in blue/white colony screening.
It inactivates the lac repressor and induces synthesis of beta-galactosidase, an enzyme that promotes lactose utilization. IPTG is used to induce expression of cloned genes under control of the lac operon. It is used in conjunction with X-Gal to determine the lac phenotype in blue/white colony screening.Ready-to-use 100mM IPTG aqueous solution is non-toxic, stable, 0.22µm membrane-filtered, formulated for direct use in blue/white colony screening.
Specifications
direct use in blue/white colony screening. | |
IPTG, dioxane-free | |
1g |
Promotions
-
Save Over 90% on Life Sciences Products
Take advantage of amazing deals while they last.: 12-31-2019
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.