Learn More
Thermo Scientific™ Maxima Probe qPCR Master Mixes
Optimize qPCR with probe chemistry using standard cycling protocols with these ready-to-use qPCR master mixes.
Manufacturer:  Thermo Scientific™ K0232
 View more versions of this product
Catalog No. FERK0232
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Maxima Probe qPCR Master Mixes are ready-to-use solutions optimized for quantitative real-time PCR. The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added.
Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG). The use of Maxima Probe qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates.
Maxima Probe qPCR Master Mixes are compatible with most real-time thermal cyclers.
The master mix includes Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. It is supplemented with ROX passive reference dye. Only template and primers need to be added.Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carryover contamination control using uracil DNA glycosylase (UDG). ROX serves as a passive reference dye. The use of Maxima Probe/ROX qPCR Master Mix (2X) in real time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral and cDNA templates. The master mix can be used with most real-time thermal cyclers, including instruments from Applied Biosystems, Eppendorf, Corbett, Bio-Rad and Roche.
  • Sensitivity—detects low copy number targets
  • Specificity—Maxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
  • Wide linear range—accurate quantification across 9 orders of magnitude
  • Universal—can be used with sequence-specific probes on most real-time thermal cyclers
  • Reproducibility and convenience—ready-to-use 2X master mix minimizes pipetting error and reduces set-up time
Recommended for:
Real-time PCR using sequence-specific probes; Real-time RT-PCR using sequence-specific probes.
Specifications
Specifications
Maxima™ Probe/ROX qPCR Master Mix (2X)
Real-time PCR using sequence-specific probes; Real-time RT-PCR using sequence-specific probes.
1000 reactions of 25 µl
10 x 1.25mL
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
† Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.