Learn More
Thermo Scientific™ NBT
Enhance blue color development during detection of alkaline phosphatase activity with NBT, an oxidant used in conjunction with BCIP or BCIP derivatives (BCIP T).
Manufacturer:  Thermo Scientific™ R0841
 View more versions of this product
Catalog No. FERR0841
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
NBT is used to enhance blue color development during detection of alkaline phosphatase activity . It is suitable for immunohistochemistry and cytochemistry. NBT is also a substrate for oxidases and dehydrogenases.
It is suitable for immunohistochemistry and cytochemistry. NBT is also a substrate for oxidases and dehydrogenases.
Recommended for:
Detection of alkaline phosphatase activity in:
  • Southern blotting
  • Western blotting
  • Northern blotting
  • dot/slot blots
  • plaque and colony screening
Note:
Preparation of a 75mg/mL stock solution in 70% dimethylformamide is recommended.
Specifications
Specifications
Detection of alkaline phosphatase activity in:; Southern blotting Western blotting Northern blotting dot/slot blots plaque and colony screening
NBT
1g