Learn More
Thermo Scientific™ NBT
Enhance blue color development during detection of alkaline phosphatase activity with NBT, an oxidant used in conjunction with BCIP or BCIP derivatives (BCIP T).
Specifications
For Use With (Application) Detection of alkaline phosphatase activity in:; Southern blotting Western blotting Northern blotting dot/slot blots plaque and colony screening
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FERR0841
View Documents
Thermo Scientific™
R0841
1g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
FERR0842
View Documents
Thermo Scientific™
R0842
5g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
NBT is used to enhance blue color development during detection of alkaline phosphatase activity . It is suitable for immunohistochemistry and cytochemistry. NBT is also a substrate for oxidases and dehydrogenases.
It is suitable for immunohistochemistry and cytochemistry. NBT is also a substrate for oxidases and dehydrogenases.
Recommended for:
Detection of alkaline phosphatase activity in:
  • Southern blotting
  • Western blotting
  • Northern blotting
  • dot/slot blots
  • plaque and colony screening
Note:
Preparation of a 75mg/mL stock solution in 70% dimethylformamide is recommended.
Specifications
Specifications
Detection of alkaline phosphatase activity in:; Southern blotting Western blotting Northern blotting dot/slot blots plaque and colony screening