Learn More
Thermo Scientific™ Primers for cDNA Synthesis
Optimze cDNA synthesis with these random hexamer primers, oligo(dT)18 primers and anchored oligo dT primer.
Specifications
For Use With (Application) cDNA Synthesis
View More Specs
Products 1
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
AB-1247
View Documents
Thermo Scientific™
AB1247
Anchored Oligo dT 100µL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Random Hexamer Primer is a mixture of single-stranded random hexanucleotides with 5'- and 3'-hydroxyl ends. The primer is supplied as a ready-to-use, 20X concentrated aqueous solution. Oligo(dT)18 Primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends. The primer is supplied as a ready-to-use, 20X concentrated aqueous solution.
Anchored Oligo-dT primers are specifically designed for synthesising long cDNAs from mRNAs carrying poly-A tails. Since this primer only anneals to the poly-A tail junction of the mRNA template, it eliminates transcription through the poly-A tail, providing more effective cDNA synthesis.
Thermo Scientific PCR reagents meet high quality standards to generate accurate results from even the most challenging molecular biology applications. Since this primer only anneals to the poly-A tail junction of the mRNA template, it eliminates transcription through the poly-A tail, providing more effective cDNA synthesis.
Recommended for:
cDNA Synthesis
Specifications
Specifications
cDNA Synthesis