Learn More
Thermo Scientific™ T3 RNA Polymerase
Catalyze the 5'→3' synthesis of RNA on either single-stranded or double-stranded DNA with T73 RNA polymerase.
Manufacturer:  Thermo Scientific™ EP0102
 View more versions of this product
Catalog No. FEREP0102
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Bacteriophage T3 RNA polymerase is DNA-dependent RNA polymerase with strict specificity for their respective double-stranded promoters. It catalyzes the 5'→3' synthesis of RNA on either single-stranded DNA or double-stranded DNA downstream from it promoter and is able to incorporate modified nucleotide.>/p>
It catalyzes the 5'=>3' synthesis of RNA on either single-stranded DNA or double-stranded DNA downstream from its promoter.
  • Incorporates modified nucleotides (e.g., aminoallyl-, biotin-, fluorescein-, digoxigenin-labeled nucleotides)
Recommended for:
Synthesis of unlabeled and labeled RNA that can be used: for hybridization (1), in vitro RNA translation (2); as aRNA (3), siRNA (4), substrate in RNase protection assays (5), template for genomic DNA sequencing.
Note:
Consensus promoter sequences: AATTAACCCTCACTAAA G GGAGA–T3 TAATACGACTCACTATA G GGAGA–T7 ATTTAGGTGACACTATA G AAGNG–SP6 The position in bold (+1) indicates the first nucleotide incorporated into RNA during transcription. Only bases at this position through +3 are critical for transcription, and they must be a G and a purine base, respectively (9).
Specifications
Specifications
20U/µL
Synthesis of unlabeled and labeled RNA that can be used: for hybridization (1), in vitro RNA translation (2); as aRNA (3), siRNA (4), substrate in RNase protection assays (5), template for genomic DNA sequencing.
10,000U
T3 RNA Polymerase
1mL of 5X Transcription Buffer