Learn More
Edvotek™ Fermentation and Bioprocessing of Chromogenic Proteins
SDPSCI_ED
Designed to introduce the basic concepts of fermentation and bioprocessing through the production of GFP protein in a small-scale fermentor. Edvotek™ Fermentation and Bioprocessing of GFP Experiment Kit is for 10 lab groups.
Supplier:  Edvotek™ 305
Catalog No. S29016
Includes: instructions, BactoBeads™ with purple and pink plasmid, LB Growth Media, Ampicillin, IPTG, protein extraction buffer, wash buffer, elution buffer, dry ion exchange matrix, standard protein markers, 50% glycerol solution, protein denaturing solution, Microcentrifuge Tubes, pH paper, centrifuge tubes, chromatography columns, protein Protein InstaStain™, practice gel loading solution, tris-glycine-SDS electrophoresis buffer, transfer pipets, loops
Requires : Air pump, Heater, pH Probe, Temperature Probe, Fermentor Vessel, Autoclave, Ice, Shaker, Ethanol, DH2O, Centrifuge, Colorimeter, Balance.
Description
Description
Bioprocessing is the production and isolation of desired products from living cells. In this introduction to bioprocessing, students will use small-scale fermenters to produce chromogenic proteins using Escherichia coli. Protein extracts will then be separated using column chromatography to analyze the success of the fermentation process. Finally, the protein solutions will be examined by SDS polyacrylamide gel electrophoresis to determine the purity of the chromogenic proteins.
Specifications
Specifications
Fermentation and Bioprocessing of GFP Experiment Kit
Complete in 5-6 hours
10 Lab Groups
Instructions, BactoBeads™ with GFP plasmid, LB Growth Media, Growth Additive, Ampicillin, IPTG, GFP Extraction Buffer, Microcentrifuge Tubes, Syringe, 15 ml Centrifuge tubes, transfer pipettes, loops.
Refrigerator and Freezer Storage (some components)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.