Learn More
FERMT1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 222151AP
Catalog No. 50-173-3775
Add to cart
Description
Description
Due to a concerted effort to identify biomarkers for lung and colon carcinomas by genome-wide transcriptional profiling, the identification and cloning of one such gene as well as two additional closely related genes was achieved. Due to the strong sequence homology to the C. elegans UNC-112 a novel gene was named URP1, for UNC-112 related protein. Another novel related gene, URP2 and the previously discovered MIG-2 gene was also identified. Transcriptional analysis shows that only URP1 is significantly differentially regulated, being over-expressed in 70% of the colon carcinomas and 60% of the lung carcinomas tested. Quantification of URP1 expression by qRT-PCR showed up-regulation of the gene by 60-fold in lung tumors and up to nearly 6-fold in colon tumors. Northern blot analysis of URP1 indicates that normal expression is restricted to neuromuscular tissues. In contrast, the expression of URP2 appears to be confined primarily to tissues of the immune system. URP1 has the potential to be one of the most important prognostic and diagnostic markers of colon or lung cancer.
Specifications
Specifications
FERMT1
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P59113, Q9BQL6
C20orf42, DTGCU2, Fermitin family homolog 1, FERMT1, KIND1, Kindlerin, Kindlin 1, Kindlin syndrome protein, Unc 112 related protein 1, UNC112A, URP1
Rabbit
IgG
150 μL
-20°C
Primary
241639, 55612
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FERMT1
Liquid
FERMT1
FERMT1 Fusion Protein Ag17543
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse