Ferno™ Cardboard Splints

Designed for splinting upper and lower extremities

Manufacturer: FERNO WASHINGTON SP1512

 View more versions of this product

Catalog No. 19-130-5675

  Check Availability
Add to Cart

    Specifications

    Color White
    For Use With (Equipment) X-ray translucent and MRI
    Length (English) 18 in.
    Length (Metric) 457.2mm
    Material Cardboard
    Product Type Splint

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit