Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Ammonium Citrate, Green, Powder, FCC, 14.5-16%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
1185-57-5
C6H8FeNO7
261.98
FRHBOQMZUOWXQL-UHFFFAOYSA-K
iron(3+) 2-hydroxypropane-1,2,3-tricarboxylate amine
N.[Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
1185-57-5
C6H8FeNO7
261.98
FRHBOQMZUOWXQL-UHFFFAOYSA-K
iron(3+) 2-hydroxypropane-1,2,3-tricarboxylate amine
N.[Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

F1001, 1185-57-5

Spectrum™ Ferric Ammonium Citrate, Green, Powder, FCC is also known as Ferric ammonium citrate or Ammonium Iron(III) Citrate and is used as an acidity regulator. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Percent Purity 14.5 to 16%
Lead (Pb) 2 mg/kg
Mercury (Hg) 1 mg/kg
Packaging Poly Pail
Sulfate 0.003
Quantity 2.5 kg