Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Chloride, 0.025% (w/v) Solution, For BOD, APHA, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly Bottle
10025-77-1
10025-77-1
Grade Chemical Solution
Packaging Poly Bottle
Quantity 500 mL