Ferric Chloride, 10% (w/v) Aqueous Solution, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 312032

Packaging Poly natural
Quantity 1L

 View more versions of this product

Catalog No. 3120-32

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7732-18-5,10025-77-1,7647-01-0
    CAS Min % 88.0,8.82,1.04
    CAS Max % 92.0,9.18,1.08
    Chemical Name or Material Ferric Chloride Solution

    Specifications

    Quantity 1L
    CAS Min % 88.0,8.82,1.04
    Packaging Poly natural
    CAS 7647-01-0,7732-18-5,10025-77-1
    CAS Max % 1.08,92.0,9.18
    Concentration or Composition (by Analyte or Components) 10% (w/v) Aqueous Solution
    Chemical Name or Material Ferric Chloride Solution

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit