Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Chloride, 25% w/v, Reagents

Catalog No. 01335297
Change view
Click to view available options
:
Quantity: 10 L
Catalog No.
01-335-297 Quantity: 10 L
1 options

Catalog No. 01-335-297

Supplier: Reagents Holdings Llc CF00170010F

Due to product restrictions, please to purchase or view availability for this product.

Ferric Chloride, 25% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10025-77-1 , 7732-18-5
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Synonym Iron(III) Chloride
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Boiling Point 102°C
Color Amber
Physical Form Liquid
Quantity 10 L
Percent Purity 25%
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.1111
Chemical Name or Material Ferric Chloride, 25% w/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.