Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Chloride, 41 DEG Be Solution, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
55 gal
Packaging:
Drum
Poly Bottle
Poly Drum
10025-77-1
10025-77-1
Grade Reagent
Packaging Poly Drum
Quantity 20 L

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.