Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Chloride, Hexahydrate, Lump, ACS, 97.0-102.0%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Poly Drum
Poly Pail
10025-77-1
10025-77-1
Grade ACS
CAS Max % 1
Percent Purity 97 to 102%
Calcium (Ca) 0.0001
Copper (Cu) 3e-05
Insoluble Matter 0.0001
Magnesium (Mg) 5e-05
Nitrate 0.0001
Packaging Amber Glass Bottle
Potassium (K) 5e-05
Sodium (Na) 0.0005
Sulfate 0.0001
Zinc (Zn) 3e-05
Quantity 500 g
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.