Ferric Chloride, T.S., BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Manufacturer: J.T.Baker 592101

Packaging Narrow Mouth Amber Glass Bottle
Quantity 500mL

 View more versions of this product

Catalog No. 02-002-862

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    Chemical Name or Material Ferric Chloride, T.S.
    Grade BAKER ANALYZED™ Reagent

    Specifications

    Packaging Narrow Mouth Amber Glass Bottle
    Specific Gravity 1L = 1.00kg
    Quantity 500mL
    Chemical Name or Material Ferric Chloride, T.S.
    Grade BAKER ANALYZED™ Reagent

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit