Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Chloride, T.S., BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
100mL
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

10025-77-1

High quality chemicals for laboratory and specialized industrial use

Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.00kg
Quantity 100mL
Chemical Name or Material Ferric Chloride, T.S.
Grade BAKER ANALYZED™ Reagent