Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Nitrate, 9-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
7782-61-8
FeH18N3O18
403.992
SZQUEWJRBJDHSM-UHFFFAOYSA-N
ferric nitrate nonahydrate,iron iii nitrate nonahydrate,unii-vmx4uop3vn,ferric nitrate, nonahydrate,iron iii nitrate,vmx4uop3vn,iron, reference standard solution,ferricnitratenonahydrate,nitric acid, iron 3+ salt, nonahydrate,iron iii nitrate hydrate
16211566
iron(3+);trinitrate;nonahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Fe+3]
7782-61-8
FeH18N3O18
403.992
SZQUEWJRBJDHSM-UHFFFAOYSA-N
ferric nitrate nonahydrate,iron iii nitrate nonahydrate,unii-vmx4uop3vn,ferric nitrate, nonahydrate,iron iii nitrate,vmx4uop3vn,iron, reference standard solution,ferricnitratenonahydrate,nitric acid, iron 3+ salt, nonahydrate,iron iii nitrate hydrate
16211566
iron(3+);trinitrate;nonahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Fe+3]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 404
Physical Form Crystals
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.68kg
Quantity 2.5kg
Chemical Name or Material Ferric Nitrate, 9-Hydrate