Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Nitrate Nonahydrate, ACS Reagent (Iron (III) Nitrate Nonahydrate), Reagents

Catalog No. p-200021094
Click to view available options
Quantity:
125 g
500 g
2.5 kg
50 kg

Ferric Nitrate Nonahydrate, ACS Reagent (Iron (III) Nitrate Nonahydrate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7782-61-8
Molecular Formula FeH18N3O18
Molecular Weight (g/mol) 403.99
MDL Number MFCD00149708
InChI Key SZQUEWJRBJDHSM-UHFFFAOYSA-N
Synonym Iron(III) Nitrate Nonahydrate
IUPAC Name iron(3+) nonahydrate trinitrate
SMILES O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 125°C
Melting Point 47.2°C
Color Pale Mauve
Physical Form Solid
Quantity 125 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.684
Chemical Name or Material Ferric Nitrate Nonahydrate, ACS Reagent (Iron (III) Nitrate Nonahydrate)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.