Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Nitrate, Nonahydrate, Crystal, ACS, 98.0-101.0%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7782-61-8
7782-61-8
Grade ACS
CAS Max % 1
Percent Purity 98 to 101%
Calcium (Ca) 0.0001
Chloride 5 ppm
Insoluble Matter 5e-05
Magnesium (Mg) 5e-05
Packaging Poly Pail
Potassium (K) 5e-05
Sodium (Na) 0.0005
Sulfate 0.0001
Quantity 12 kg
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.