Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Oxide, Powder, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle

Chemical Identifiers

1309-37-1
Fe2O3

High quality chemicals for laboratory and specialized industrial use

CAS 1309-37-1
Molecular Formula Fe2O3
Formula Weight 159.69
Physical Form Powder
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 5.24kg
Quantity 500g
Chemical Name or Material Ferric Oxide
Grade BAKER ANALYZED™ Reagent