Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Subsulfate Solution, USP, 20-22 g/100 mL, Spectrum™

Click to view available options
Quantity:
500 mL
6 x 500 mL
200 L
Packaging:
Amber Glass Bottle
Drum
1310-45-8
Fe4H2O22S5
737.67
GDPKWKCLDUOTMP-UHFFFAOYSA-B
tetrairon(3+) dihydroxide pentasulfate
[OH-].[OH-].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
1310-45-8
Fe4H2O22S5
737.67
GDPKWKCLDUOTMP-UHFFFAOYSA-B
tetrairon(3+) dihydroxide pentasulfate
[OH-].[OH-].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

F1305, 1310-45-8, Fe4(OH)2(SO4)5

Spectrum™ Ferric Subsulfate Solution, USP All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Concentration 20-22 g/100 mL
Packaging Amber Glass Bottle
Quantity 6 x 500 mL