Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Sulfate, n-Hydrate, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle

Chemical Identifiers

15244-10-7
Fe2H2O13S3
417.873
VXWSFRMTBJZULV-UHFFFAOYSA-H
167265

High quality chemicals for laboratory and specialized industrial use

CAS 15244-10-7
Molecular Formula Fe2H2O13S3
InChI Key VXWSFRMTBJZULV-UHFFFAOYSA-H
SMILES O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3]
IUPAC Name iron(3+);trisulfate;hydrate
Molecular Weight (g/mol) 417.873
PubChem CID 167265
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 3.10kg
Quantity 500g
Chemical Name or Material Ferric Sulfate, n-Hydrate
Grade BAKER ANALYZED™ Reagent
Show More Show Less