Learn More
Drg International IncSupplier Diversity Partner Ferritin Elisa DFS Item
Manufacturer:  Drg International IncSupplier Diversity Partner EIA1872
An enzyme immunoassay for the quantitative determination of ferritin concentration in serum. One of the most prevalent disorders of man is the dietary deficiency of iron and the resulting anemia. Serum ferritin levels can be measured routinely and are particularly useful in the early detection of iron-deficiency anemia in apparently healthy people. The Ferritin Enzyme Immunoassay provides a rapid, sensitive, and reliable assay. The assay system utilizes rabbit anti-ferritin for the solid phase (microtiter wells) immobilization and mouse monoclonal anti-ferritin in the antibody-enzyme (horseradish peroxidase) conjugate solution.
Catalog No. 50-194-7546