Learn More
Abnova™ Ferritin (Human) ELISA Kit
Sandwich enzyme immunoassay for the quantitative measurement of human Ferritin
Manufacturer:  Abnova™ KA4793
Catalog No. 89-944-394
Add to cart
Description
Description
  • Used for the in vitro quantitative determination of human Ferritin in human serum, human plasma, cell lysate and buffered solution
  • Assay will recognize native Ferritin
Specifications
Specifications
P02792;P02794
Biotin
Ferritin (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human Ferritin.
2512
ELISA Kit
Buffered solution, Cell lysate, Plasma, Serum
Store the kit at 4°C.
0.23 to 15 ng/mL
Human
Quantification
AV-HRP
For research use only (RUO)
100μL
Colorimetric