Learn More
Abnova™ Ferritin (Human) ELISA Kit
Sandwich enzyme immunoassay for the quantitative measurement of ferritin.
Manufacturer:  Abnova™ KA4821
Catalog No. 89-032-267
Add to cart
Description
Description
  • For the quantitative determination of Human Ferritin concentration in human serum
Specifications
Specifications
0 to 1000 ng/mL
Ferritin (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of ferritin.
Antibody coated microtiter plate
For research use only (RUO)
Store the kit at 4°C.
Human
HRP
Quantification
ELISA Kit
20 μL
Colorimetric
Serum