Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferritin Light Chain Mouse anti-Human, Allophycocyanin/Cy7, Clone: FTL/1389, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254583APCCY7

Catalog No. N254583APC7


Add to cart

Description

Description

Ferritin Light Chain Monoclonal antibody specifically detects Ferritin Light Chain in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Ferritin Light Chain
FTL/1389
Flow Cytometry
Conjugated, Purified
FTL
Recombinant human FTL protein fragment (aa 38-165) (exact sequence is proprietary)
Protein A or G purified
Cancer
Primary
2512.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
Ferritin L subunit, ferritin L-chain, ferritin light chain, ferritin light polypeptide-like 3, ferritin, light polypeptide, MGC71996, NBIA3
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit