Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferritin Mouse anti-Human, Clone: FTH/2081, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Ferritin
Clone FTH/2081
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 0.1-0.2 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123064
View Documents
Novus Biologicals
NBP30779520UG
20 μg
Each for $138.37
Add to cart
 
NB123063
View Documents
Novus Biologicals
NBP30779510UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

Ferritin Monoclonal antibody specifically detects Ferritin in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications

Specifications

Ferritin
0.2 mg/ml
Western Blot, Immunohistochemistry (Paraffin)
Purified
RUO
Human
apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15
Recombinant human Ferritin protein fragment (around aa 58-180) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
FTH/2081
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 0.1-0.2 ug/ml
Unconjugated
Mouse
Cancer, Cellular Markers, Lipid and Metabolism
10 mM PBS with 0.05% BSA
2495
IgG2c Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit