Learn More
Ferritin Mouse anti-Human, DyLight 550, Clone: F31, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB1108385R
Catalog No. NB1108385R
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Ferritin Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Enzyme Linked Immunosorbent Assay.
Specifications
Specifications
Ferritin
F31
ELISA, Sandwich ELISA
Purified
FTH1
This clone has been derived from hybridization of SP2/0 myeloma cells with spleen cells of Balb/c mice immunised with purified human ferritin.
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Ferritin (F31)
ELISA
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15
Mouse
IgG2b
0.2 ml
Cancer, Cellular Markers, Lipid and Metabolism
Primary
2495
Human