Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferroceneacetic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
1287-16-7
C12H12FeO2
MFCD00045464
1287-16-7
C12H12FeO2
MFCD00045464
Quantity 1g
Packaging Glass bottle
Color Brown-Orange to Golden
Melting Point 156°C to 160°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Formula Weight 244.07
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material Ferroceneacetic acid

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.