Alfa Aesar™ Ferrocenecarboxaldehyde, 97%

Manufacturer: Alfa Aesar™ 3925409

Quantity 10g

 View more versions of this product

Catalog No. AA3925409

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12093-10-6
    Formula Weight 214.05
    MDL Number MFCD00001429
    Synonym [Cyclopentadienyl(formylcyclopentadienyl)iron]
    Chemical Name or Material Ferrocenecarboxaldehyde

    Specifications

    Formula Weight 214.05
    MDL Number MFCD00001429
    Physical Form Crystalline
    CAS 12093-10-6
    Sensitivity Air Sensitive
    Melting Point 124°C
    Quantity 10g
    TSCA TSCA
    Synonym [Cyclopentadienyl(formylcyclopentadienyl)iron]
    Chemical Name or Material Ferrocenecarboxaldehyde
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit