Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrocenecarboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-3757928
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
12093-10-6
C11H10FeO
214.05
MFCD00001429
UQTCQJVPLIVCAX-UHFFFAOYNA-N
Cyclopentadienyl(formylcyclopentadienyl)iron
(cyclopenta-2,4-dien-1-yl)(2-formylcyclopenta-2,4-dien-1-yl)iron
[Fe].c1cccc1.O=Cc1cccc1
12093-10-6
C11H10FeO
214.05
MFCD00001429
UQTCQJVPLIVCAX-UHFFFAOYNA-N
Cyclopentadienyl(formylcyclopentadienyl)iron
(cyclopenta-2,4-dien-1-yl)(2-formylcyclopenta-2,4-dien-1-yl)iron
[Fe].c1cccc1.O=Cc1cccc1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
TRUSTED_SUSTAINABILITY
Melting Point 120.0°C to 124.0°C
Color Brown to Orange
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 214.05
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Ferrocenecarboxaldehyde
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit