Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrocenemethanol, 97%, ACROS Organics™

Click to view available options
Quantity:
500mg
Packaging:
Glass bottle
1273-86-5
C11H12FeO
MFCD00040878
1273-86-5
C11H12FeO
MFCD00040878
Quantity 500mg
Infrared Spectrum Authentic
Color Orange to Yellow
Packaging Glass bottle
Melting Point 77°C to 82°C
Assay Percent Range 97%
Formula Weight 215.93
Physical Form Powder or Needles
Percent Purity ≥96.0%
Chemical Name or Material Ferrocenemethanol

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.