Alfa Aesar™ (Ferrocenylmethyl)trimethylammonium iodide

Manufacturer: Alfa Aesar™ 3939906

Quantity 5g

 View more versions of this product

Catalog No. AA3939906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12086-40-7
    Formula Weight 385.08
    MDL Number MFCD00041534
    Synonym N,N-Dimethylaminomethylferrocene methiodide
    Chemical Name or Material (Ferrocenylmethyl)trimethylammonium iodide

    Specifications

    Formula Weight 385.08
    Physical Form Crystalline
    MDL Number MFCD00041534
    CAS 12086-40-7
    Melting Point 220°C (dec.)
    Synonym N,N-Dimethylaminomethylferrocene methiodide
    Quantity 5g
    Chemical Name or Material (Ferrocenylmethyl)trimethylammonium iodide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit