Learn More
Ferroin Indicator, 0.025 M Phenanthroline Ferrous Sulfate Complex, Ricca Chemical
Supplier: Ricca Chemical Company 31404
Specifications
Ferroin Indicator | |
96.0, 1.43, 0.67 | |
5144-89-8, 7782-63-0 | |
Amber Glass Bottle | |
120 mL |
Phenanthroline Ferrous Sulfate Complex | |
100.0, 1.48, 0.69 | |
0.025Molar | |
Red | |
Liquid |