Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ferroin Indicator Solution, 0.025M Orthophenanthroline TS, Reagents
Click to view available options
Quantity:
100 mL
500 mL
1 L
Specifications
Specifications
Chemical Name or Material | Ferroin Indicator Solution, 0.025M Orthophenanthroline TS |
CAS | 7732-18-5, 5144-89-8, 5144-89-8 |
Boiling Point | 100°C |
Synonym | O-Phenanthroline, 1,10-Phenanthroline |
Grade | Reagent |
Packaging | Glass Bottle |
Specific Gravity | 1.01 |
Color | Dark Red |
Quantity | 1 L |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction