Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ferroportin/SLC40A1 Rabbit anti-Human, Alexa Fluor 750, Clone: 1308C, R&D Systems™
Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB9924S100UG
This item is not returnable.
View return policy
Description
Ferroportin/SLC40A1 Monoclonal specifically detects Ferroportin/SLC40A1 in Human samples. It is validated for Flow Cytometry.Specifications
Ferroportin/SLC40A1 | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Ferroportin-1, FPN1, FPN1IREG1ferroportin 1, HFE4, HFE4ferroportin-1, IREG1, iron regulated gene 1, Iron-regulated transporter 1, member 3, MST079, MSTP079, MTP1, putative ferroportin 1 variant IIIB, SLC11A3, SLC11A3iron regulated gene 1, SLC40A1, solute carrier family 11 (proton-coupled divalent metal ion transporters), solute carrier family 11 (proton-coupled divalent metal ion transporters), member 3, solute carrier family 40 (iron-regulated transporter), member 1, solute carrier family 40 member 1 | |
Synthetic peptide containing human Ferroportin/SLC40A1, Accession No. Q9NP59 | |
100 μg | |
30061.0 | |
Human | |
Supernatant |
Flow Cytometry | |
1308C | |
Flow Cytometry 0.25-1 μg/106 cells | |
Q9NP59 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human Ferroportin/SLC40A1 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title