Alfa Aesar™ Ferrosilicon 75

Manufacturer: Alfa Aesar™ 4554722

Quantity 100g

 View more versions of this product

Catalog No. AA4554722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 8049-17-0
    MDL Number MFCD00016087
    UN Number UN1408
    Chemical Name or Material Ferrosilicon 75

    Specifications

    Melting Point >1000°C
    Quantity 100g
    Health Hazard 3 GHS P Statement
    P231+P232-P261-P280-P304+P340-P405-P501a
    Handle under inert gas.
    Protect from moisture.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H331-H261-H302-H312
    Toxic if inhaled.
    In contact with water releases flammable gas.
    Harmful if swallowed.
    Harmful in contact with skin.
    UN Number UN1408
    Concentration ≈75% silicon
    Color Gray
    Physical Form Granules
    MDL Number MFCD00016087
    CAS 8049-17-0
    Health Hazard 1 Danger
    RTECS Number LK1400000
    DOT Information Transport Hazard Class: 4.3; Packing Group: III; Proper Shipping Name: FERROSILICON
    Chemical Name or Material Ferrosilicon 75

    GHS H Statement
    H331-H261-H302-H312
    Toxic if inhaled.
    In contact with water releases flammable gas.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P231+P232-P261-P280-P304+P340-P405-P501a
    Handle under inert gas.
    Protect from moisture.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit