Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrosoferric Oxide, Black, Powder, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
1317-61-9
Fe3O4
231.53
SZVJSHCCFOBDDC-UHFFFAOYSA-N
oxo[(oxoferrio)oxy]iron; oxoiron
O=[Fe].O=[Fe]O[Fe]=O
1317-61-9
Fe3O4
231.53
SZVJSHCCFOBDDC-UHFFFAOYSA-N
oxo[(oxoferrio)oxy]iron; oxoiron
O=[Fe].O=[Fe]O[Fe]=O

F1058, 1317-61-9, Fe3O4

Spectrum™ Ferrosoferric Oxide, Black, Powder
CAS Max % 1
Packaging Amber Glass Bottle
Quantity 2.5 kg