Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ferrous Ammonium Sulfate, 0.1 Normal (N/10), Unstandardized, Reagents
Click to view available options
Quantity: 10 L
Specifications
Specifications
Chemical Name or Material | Ferrous Ammonium Sulfate, 0.1 Normal (N/10), Unstandardized |
Boiling Point | 100°C |
CAS | 7732-18-5, 7783-85-9 |
Color | Light Green |
Physical Form | Liquid |
Quantity | 10 L |
Synonym | Iron(II) Ammonium Sulfate |
Grade | Reagent |
Packaging | Cubitainer |
Specific Gravity | 1.05 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction