Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Ammonium Sulfate, 0.1N Mohr's Salt Solution, Reagents

Catalog No. 01334710
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Catalog No. Quantity
01-334-710 20 L
01-334-711 1 L
01-334-709 4 L
01-334-708 500 mL
4 options

Catalog No. 01-334-710

Supplier: Reagents Holdings Llc CM12970020F

Only null left
Add to Cart
Add to Cart

Ferrous Ammonium Sulfate, 0.1N Mohr's Salt Solution, Requires Periodic Restandardization, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Specifications

Boiling Point 100°C
Chemical Name or Material Ferrous Ammonium Sulfate, 0.1N Mohr's Salt Solution, Requires Periodic Restandardization
Color Light Green
CAS 7732-18-5, 7664-93-9
pH <1
Physical Form Liquid
Quantity 20 L
Synonym Iron(II) Ammonium Sulfate
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.05
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.